ContactUsLinkComponent
2X SDS-PAGE Sample Buffer, Rockland Immunochemicals
Catalog # 10799-488
CAS Number:  
2X SDS-PAGE Sample Buffer, Rockland Immunochemicals
Catalog # 10799-488
Supplier Number:  KBB-001
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    2X SDS-PAGE Sample Buffer
  • Size:
    1.5 mL
  • Environmentally Preferable:
  • Shipping Temperature:
    Dry Ice
  • Cat. No.:
    10799-488
  • Supplier no.:
    KBB-001
Specifications
About this item
2X SDS-PAGE Sample Buffer
  • 0.125 M Tris-HCl, 4% (w/v) SDS, 20% (v/v) Glycerol and 0.01% (w/v) bromphenol blue, pH6.8
  • aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers
  • The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
SDS-PAGE, sodium dodecyl sulfate polyacrylamide gel electrophoresis, describes a technique widely used in biochemistry to separate proteins according to their electrophoretic mobility (a function of the length of a polypeptide chain and its charge) and no other physical feature. SDS is an anionic detergent applied to protein sample to linearize proteins and to impart a negative charge to linearized proteins.
2X SDS-PAGE Sample Loading Buffer is suitable for laboratory involved in protein biochemistry. Visit our newly expanded web site at www.rockland-inc.com for methods using this and other buffers.
This product is a concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration. 2X SDS-PAGE Sample Buffer consists of 0.125 M Tris-HCl, 4% (w/v) SDS, 20% (v/v) Glycerol and 0.01% (w/v) bromphenol blue.  The pH of this solution is 6.8.  Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionized water.