ContactUsLinkComponent
Monkey (Cynomolgus) Plasma Sterile In Sodium Citrate, Rockland Immunochemicals
Monkey (Cynomolgus) Plasma Sterile In Sodium Citrate, Rockland Immunochemicals
Catalog # 75929-994
Monkey (Cynomolgus) Plasma Sterile In Sodium Citrate, Rockland Immunochemicals
Catalog # 75929-994
Supplier Number:  D516-03-2
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Monkey (Cynomolgus) Plasma Sterile In Sodium Citrate
  • Size:
    10 mL
  • Environmentally Preferable:
  • Shipping Temperature:
    Dry Ice
  • Cat. No.:
    75929-994
  • Supplier no.:
    D516-03-2
Specifications
About this item
Sterile Plasma in Anticoagulant
  • 0.22 micron filter and found to be negative for bacterial growth after incubation on trypticase soy agar for 24 hours, 48 hours, and 72 hours
  • prepared by collection of whole blood into said anticoagulant
  • Red blood cells are removed through a process of centrifugation and filtration techniques - leaving non-sterile plasma behind
used as a component of bioassays, immunoassays or enzyme assays
used as a supplement to cell culture media
provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health
Be certain to maintain Good Cell Culture Practice, and maintain sterility of cultures that require media supplementation.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal