ContactUsLinkComponent
Pigeon Plasma Non-Sterile In Sodium EDTA, Rockland Immunochemicals
Pigeon Plasma Non-Sterile In Sodium EDTA, Rockland Immunochemicals
  75930-964
Pigeon Plasma Non-Sterile In Sodium EDTA, Rockland Immunochemicals
  75930-964
 :  D412-04
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Pigeon Plasma Non-Sterile In Sodium EDTA
  • Size:
    10 mL
  • Environmentally Preferable:
  • Shipping Temperature:
    Dry Ice
  • Cat. No.:
    75930-964
  • Supplier no.:
    D412-04
Non-Sterile Plasma in Anticoagulant
  • prepared by collection of whole blood into said anticoagulant
  • Red blood cells are removed through a process of centrifugation and filtration techniques - leaving non-sterile plasma behind
used as a component of bioassays, immunoassays or enzyme assays
provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health
also available as a sterile preparation for use as a cell culture supplement
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal