ContactUsLinkComponent
Horse Serum (Lyophilized), Rockland Immunochemicals
Horse Serum (Lyophilized), Rockland Immunochemicals
Catalog # 75934-582
Horse Serum (Lyophilized), Rockland Immunochemicals
Catalog # 75934-582
Supplier Number:  D307-05
Specifications
  • Description:
    Horse Serum (Lyophilized)
  • Size:
    5 mL
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    75934-582
  • Supplier no.:
    D307-05
Specifications
About this item
Lyophilized Non-Sterile Serum
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Horse Serum is used as a component to bioassays, immunoassays, or enzyme assays. Horse Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Be certain to maintain Good Cell Culture Practice, and maintain sterility of cultures that require media supplementation.
Suitable for use as a control or diluent in assays and most other immunological methods.