Order Entry
Puerto Rico
ContactUsLinkComponent
Duck Serum Sterile, Rockland Immunochemicals
Duck Serum Sterile, Rockland Immunochemicals
  10802-554
Duck Serum Sterile, Rockland Immunochemicals
  10802-554
 :  D114-00

 

  • Description:
    Duck Serum Sterile
  • Size:
    10 mL
  • Shipping Conditions:
    Dry Ice
  • Cat. No.:
    10802-554
  • Supplier no.:
    D114-00

 

 

Sterile Serum

  • 0.22 micron filter and found to be negative for bacterial growth after incubation on trypticase soy agar for 24 hours, 48 hours, and 72 hours
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques

Duck Serum is used as a supplement to cell culture media. Duck Serum is also suitable for use as a component of bioassays, immunoassays or enzyme assays. Duck Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Be certain to maintain Good Cell Culture Practice, and maintain sterility of cultures that require media supplementation. Duck Serum is ideal for investigators in Cancer, Immunology, and Cell Biology research.

pH: normal

Immunoelectrophoresis: normal

Hemoglobin: normal

IgG Concentration: normal