Goat Complement (Fresh Frozen), Rockland Immunochemicals
Supplier: Rockland Immunochemical
Certificates
About this item
Complement Fresh Frozen
- prepared by removal of fibrinogen through clot formation under proper temperature conditions
- Complement is preservative FREE
Special processing techniques are used to yield products with high complement activity and low background cytotoxicity. Goat Complement is suitable for CFT and SRH.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
Immunoelectrophoresis: normal
Specifications
- Catalog No:
- RLC309-0100
- RLC309-0050
- 10801-772
- RLC309-0500
- Shipping Conditions:
- Dry Ice
- Dry Ice
- Dry Ice
- Dry Ice