Rabbit 'B' (1-2 Lbs.) Complement (Fresh Frozen), Rockland Immunochemicals
Complement Fresh Frozen
- prepared by removal of fibrinogen through clot formation under proper temperature conditions
- Complement is preservative FREE
Special processing techniques are used to yield products with high complement activity and low background cytotoxicity.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
Immunoelectrophoresis: normal
- Catalog No:
- RLC303-0010
- RLC303-0050
- Shipping Conditions:
- Dry Ice
- Dry Ice