Order Entry
Puerto Rico
ContactUsLinkComponent
Goat Complement, Rockland Immunochemicals
Goat Complement, Rockland Immunochemicals
Catalog # RLC209-0005
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Goat Complement, Rockland Immunochemicals
Catalog # RLC209-0005
Supplier Number:  C209-0005
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Specifications

  • Description:
    Goat Complement (Lyophilized) With Diluent
  • Size:
    5 mL
  • Shipping Conditions:
    Ambient
  • Cat. No.:
    RLC209-0005
  • Supplier no.:
    C209-0005

Specifications

About this item

Complement Lyophilized with DILUENT

  • suitable for complement fixation tests (CFT) and Single radial haemolysis (SRH)

Special processing techniques are used to yield products with high complement activity and low background cytotoxicity.

pH: normal

Immunoelectrophoresis: normal

Hemoglobin: normal

IgG Concentration: normal

Immunoelectrophoresis: normal