Specifications
- Description:Rabbit ''B'' (1-2 Lbs.) Complement (Lyophilized) With Diluent
- Size:5 mL
- Shipping Conditions:Ambient
- Cat. No.:RLC203-0005
- Supplier no.:C203-0005
Specifications
About this item
Complement Lyophilized with DILUENT
- suitable for complement fixation tests (CFT) and Single radial haemolysis (SRH)
The rabbit complement system helps or “complements” the ability of antibodies and phagocytic cells to clear pathogens from an organism. Complement is part of the innate immune system that is not adaptable and does not change over the course of an individual's lifetime. Complement can be recruited and brought into action by the adaptive immune system.
The following are basic functions of complement:
1. Opsonisation - enhancing phagocytosis of antigens
2. Chemotaxis - attracting macrophages and neutrophils
3. Cell Lysis - rupturing membranes of foreign cells
4. Clumping of antigen-bearing agents
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
Immunoelectrophoresis: normal