Rat Glial Derived NTF
Recombinant Rat Glial Derived NTF control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Glial Cell Line-Derived Neurotrophic Factor (GDNF) is a neurotrophic factor that is closely related to other neurotrophic factors, such as Neurturin, Persephin, and Artemin, by a common structural feature called the cysteine-knot. GDNF signals through a multicomponent system of receptors that includes RET and GFRα1-4, to promote dopamine uptake, survival and differentiation of neurons. Recombinant rat GDNF is a non-glycosylated homodimer, containing two 135 amino acid chains, with a total molecular weight of 30.1 kDa.
Glial Derived Neurotrophic Factor Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Glial Derived Neurotrophic Factor in immunological assays.
- Species:Rat
- Protein/Peptide Name:Glial Derived NTF
- Shipping Temperature:Ambient