About this item
Recombinant Rat Leptin control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Leptin is a protein that is thought to have a critical role in the physiologic regulation of body weight via its capacity to inhibit food intake and stimulate energy expenditure. Leptin also has thermogenic actions and regulates enzymes of fatty acid oxidation. Severe hereditary obesity in rodents and humans can be caused by defects in leptin production. These functions include the regulation of hematopoiesis, angiogenesis, wound healing, inflammation, and immune responses. Recombinant rat Leptin is a non-glycosylated protein, containing 147 amino acids, with a molecular weight of 16.2 kDa.
Leptin Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Leptin in immunological assays.
Specifications
- Protein/Peptide Type:Recombinant
- Species:Rat
- Protein/Peptide Name:Leptin
- Shipping Temperature:Ambient