Rat Recombinant IL-6
Recombinant Rat IL-6 control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Interleukin 6 (IL-6) is an important pro-inflammatory and anti-inflammatory cytokine expressed by T cells, macrophages and muscle cells. IL-6 signals through a receptor complex containing two receptors, IL-6Rα and gp130. IL-6 has an important function in promoting fever and can serve to stimulate an immune response to trauma. IL-6 is often used for growth of hybridoma cell lines. Human IL-6 is active on mouse and rat cells. Recombinant rat IL-6 is a non-glycosylated protein, containing 188 amino acids, with a molecular weight of 21.8 kDa.
Interleukin-6 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-6 in immunological assays.
- Protein/Peptide Type:Recombinant
- Species:Rat
- Protein/Peptide Name:IL-6
- Shipping Temperature:Ambient