Mouse Recombinant LIF
Supplier: Rockland Immunochemical
Certificates
About this item
Recombinant Mouse LIF control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Mouse Leukemia Inhibitory Factor (LIF) is member of the IL-6 family and is made by a variety of tissues. LIF signals through receptors gp190 (LIFR) and gp130 to activate STAT3 which ultimately promotes mouse embryonic stem cell self-renewal and pluripotentcy of long term cultures. This activity is similar to that attributed to FGF-basic (FGF-2) for human embryonic stem cells. Recombinant mouse LIF is a non-glycosylated protein, containing 181 amino acids, with a molecular weight of 20 kDa.
Leukemia Inhibitory Factor Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Leukemia Inhibitory Factor in immunological assays.
Specifications
- Protein/Peptide Type:Recombinant
- Species:Mouse
- Protein/Peptide Name:LIF
- Shipping Temperature:Ambient