Mouse Recombinant SDF-1 alpha/CXCL12
Supplier: Rockland Immunochemical
Certificates
About this item
Recombinant Mouse SDF-1 alpha/CXCL12 control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Stromal Cell Derived Factor-1 alpha (SDF-1α), also called CXCL12, is one of two splice variants made by a wide variety of cells when stimulated by inflammatory cytokines such as, TNF, IL-1 or LPS. SDF-1α signals through the G protein-couple receptor, CXCR4, to recruit activated leukocytes. Human and mouse SDF-1α share 99% sequence identity. Recombinant mouse SDF-1α is a non-glycosylated protein, containing 68 amino acids, with a molecular weight of 7.9 kDa.
Stromal Cell-Derived Factor-1 alpha Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Stromal Cell-Derived Factor-1 alpha in immunological assays.
Specifications
- Species:Mouse
- Protein/Peptide Name:SDF-1 alpha/CXCL12
- Shipping Temperature:Ambient
- Protein/Peptide Type:Recombinant