Human Recombinant PDGF-BB
Supplier: Rockland Immunochemical
Certificates
About this item
Recombinant Human PDGF-BB control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Platelet-Derived Growth Factor (PDGF) is a mitogenic peptide growth hormone carried in the alpha-granules of platelets and is released when platelets adhere to traumatized tissues. Connective tissue cells near the traumatized region respond by initiating the process of replication. The synthesis of PDGF can be induced by IL-1, IL-6, TNF-α, TGF-β and EGF. Recombinant human PDGF-BB is a non-glycosylated, disulfide-linked homodimer, containingtwo 109 amino acid chains, with a total molecular weight of 24.3 kDa.
Platelet Derived Growth Factor-BB Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Platelet Derived Growth Factor-BB in immunological assays.
Specifications
- Protein/Peptide Type:Recombinant
- Species:Human
- Protein/Peptide Name:PDGF-BB
- Shipping Temperature:Ambient