- Conjugation:RPE (R-Phycoerythrin)
- Species:Human
- Size:0.5 ml
- Protein/Peptide Name:IgG PE
- Shipping Temperature:Ambient
- Cat. No.:RL009-008-002
- Supplier no.:009-008-002
HUMAN IgG whole molecule Phycoerythrin conjugated
- control reagent
- Flow Cytometry, Immunohistochemistry, and Western Blotting
Secreted as part of the adaptive immune response by plasma B cells, immunoglobulin G constitutes 75% of serum immunoglobulins. Immunoglobulin G binds to viruses, bacteria, as well as fungi and facilitates their destruction or neutralization via agglutination (and thereby immobilizing them), activation of the compliment cascade, and opsinization for phagocytosis.
The whole IgG molecule possesses both the F(c) region, recognized by high-afinity Fc receptor proteins, as well as the F(ab) region possessing the epitope-recognition site. Both heavy and light chains of the antibody molecule are present. This Human IgG whole molecule is conjugated to biotin (Vitamin H), a small biomolecule that has a large affinity for avidin and streptavidin.
Human IgG whole molecule Phycoerythrin conjugated can be utilized as a control reagent in Flow Cytometry, Immunohistochemistry, and Western Blotting. Researchers should determine optimal titers for applications that are not stated.