Human Recombinant Neuregulin 1Beta
Supplier: Rockland Immunochemical
Certificates
About this item
Recombinant Human Neuregulin 1Beta control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Neuregulin 1-beta (NRG1) is one of many isoforms of NRG that contains a soluble EGF-like domain. Alpha and beta varients are distinguished by difference in length at the C terminus. The EGF-like domain signals through receptors ErbB2, ErbB3, and ErbB4 to act as a growth factor. NRG isoforms are particularly important for nervous system and cardiovasucular development. Recombinant human NRG1-beta is a non-glycosylated protein, containing 66 amino acids, with a molecular weight of 7.6 kDa.
Neuregulin 1-beta Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Neuregulin 1-beta in immunological assays.
Specifications
- Protein/Peptide Type:Recombinant
- Species:Human
- Protein/Peptide Name:Neuregulin 1Beta
- Shipping Temperature:Ambient