Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Gro-alpha/CXCL1 (from E. coli)
Human Recombinant Gro-alpha/CXCL1 (from E. coli)
Certificates
Human Recombinant Gro-alpha/CXCL1 (from E. coli)

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Species:
    Human
  • Protein Synonyms:
    CXCL1
  • Protein/Peptide Name:
    GRO alpha
  • Shipping Temperature:
    Ambient
  • Source:
    E. coli
  • Storage Conditions:
    Store vial at 4 °C prior to restoration. Dilute only prior to immediate use. Maintain sterility. This product DOES NOT contain preservative. DO NOT VORTEX. We recommend adding a carrier protein such as HSA or BSA to 0.1% (i.e. 1.0 mg/mL). For best results aliquot contents and freeze at –20 °C or colder. Avoid cycles of freezing and thawing. Centrifuge vial before each opening to dislodge contents from the cap and to clarify if contents are not clear after standing at room temperature.
  • Endotoxin Content:
    Measured by kinetic LAL analysis and is typically ≤ 1 EU/μg protein.
  • Biological Activity:
    The activity is determined by the ability to chemoattract human neutrophils at concentrations between 10-100 ng/mL.
  • Gene ID:
    P09341
  • Reconstitution Instructions:
    Restore with deionized water (or equivalent); 5µl (5-50µl)
  • Purity:
    >98%
  • Formulation:
    Lyophilized

Specifications