About this item
Recombinant Human FGF-4 control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Fibroblast Growth Factor 4 (FGF-4) is a growth factor predominantly expressed during embryonic development, playing a key role in limb development. In culture, FGF-4 has been shown to be an important regulator of growth for stem cells, fibroblasts and endothelial cells. Unglycosylated FGF-4 is N-terminally cleaved into 13 kDa or 15 kDa proteins that are more active than the precursor 19 kDa protein, Bellosa P, et al. (1993). Human FGF-4 shares high homology and cross-reactivity with the mouse protein. Recombinant human FGF-4 is a non-glycosylated protein containing 140 amino acids, with a total molecular weight of 15 kDa.
Fibroblast Growth Factor-4 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Fibroblast Growth Factor-4 in immunological assays.
Specifications
- Protein/Peptide Type:Recombinant
- Species:Human
- Protein/Peptide Name:FGF-4
- Shipping Temperature:Ambient