About this item
SETMAR control protein-GST fusion
- Control Protein
- PCA Assays
- ImmunoBlot
SETMAR or SET domain and mariner transposase fusion gene is a nonhomologous end-joining repair protein that regulates genomic integration of exogenous DNA by opening chromatin and facilitating joining of DNA ends (1). SETMAR has histone methyltransferase activity and methylates 'Lys-4' and 'Lys-36' of histone H3. SETMAR also has DNA nicking activity and may play a role in DNA repair (2). Human Pso4 forms a stable complex with SETMAR that regulates Metnase function in DNA repair. SETMAR has sequence-specific DNA-binding activity and recognizes the 19-mer core of the 5'-terminal inverted repeats (TIRs) of the Hsmar1 element. SETMAR Protein is ideal for investigators involved in Signaling Proteins, Acetyl/Methyltransferase Proteins, Apoptosis/Autophagy, Cancer, Cell Cycle, and Neurobiology research.
SETMAR Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. SETMAR Protein is suitable for use in Western Blot. Expect a band approximately ~66-69kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Specifications
- Protein/Peptide Name:SETMAR
- Shipping Temperature:Dry Ice