Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant CSF2
Human Recombinant CSF2
Catalog # RL009-001-314
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Human Recombinant CSF2
Catalog # RL009-001-314
Supplier Number:  009-001-314
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Species:
    Human
  • Size:
    20 µg
  • Protein/Peptide Name:
    CSF2
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    RL009-001-314
  • Supplier no.:
    009-001-314

Specifications

About this item

GM-CSF Human Recombinant control protein

  • Control Protein
  • PCA Assays
  • ImmunoBlot

Granulocyte Macrophage Colony Stimulating Factor is produced in response to a number of inflammatory mediators by mesenchymal cells present in the hemopoietic environment and at peripheral sites of inflammation. Granulocyte Macrophage-CSF is able to stimulate the production of neutrophilic granulocytes, macrophages, and mixed granulocyte-macrophage colonies from bone marrow cells and can stimulate the formation of eosinophil colonies from fetal liver progenitor cells. GM-CSF can also stimulate some functional activities in mature granulocytes and macrophages. GM-CSF receptors shows significant homologies with other receptors for hematopoietic growth factors , including IL2-beta, IL-3 , IL -6 , IL-7 , EPO and the Prolactin receptors. Recombinant Human Granulocyte Macrophage CSF produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 127 amino acids and having a molecular weight of 14.5 kDa.

GM-CSF is suitable as a control for polyclonal or monoclonal anti-GM-CSF in immunological assays.