Order Entry
Puerto Rico
ContactUsLinkComponent
Histone H3, 21-44 Fluorescein conjugated, FITC (Fluorescein Isothiocyanate)
Histone H3, 21-44 Fluorescein conjugated, FITC (Fluorescein Isothiocyanate)
Catalog # RL000-002-K13
CAS Number:  
Histone H3, 21-44 Fluorescein conjugated, FITC (Fluorescein Isothiocyanate)
Catalog # RL000-002-K13
Supplier Number:  000-002-K13
CAS Number:  

Specifications

  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Size:
    1 mg
  • Protein/Peptide Name:
    Histone H3, 21-44 Fluorescein conjugated
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    RL000-002-K13
  • Supplier no.:
    000-002-K13

Specifications

About this item

Histone H3 Peptide Conjugated

  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot

The nucleosome is comprised of 146 bp of DNA wrapped around a series of histone proteins arranged as an octamer consisting of 2 copies of histone H2A, H2B, H3 and H4. Within the nucleosome core the histone proteins are covalent modified at specific residues predominantly within the N-terminal tail including lysine (acetylation, methylation, SUMOylation, and ubiquitinylation), arginine methylation and citrullination, serine and threonine phosphorylation, as well as proline isomerization. The lysine side chains can carry up to three methyl groups (mono-, di- and tri- methylated forms) and the arginine side chain can be monomethylated or can be dimethylated as the symmetric or asymmetric forms. The modifications show temporal, disease-specific, and other types of cell-specific regulation and there are specific families of enzymes that regulate the methylation, demethylation, acetylation, deacetylation and other modifications. Research has indicated that whereas the histone mark H3K4Me3 (tri-methyl lysine 4 of histone H3) localizes to gene promoter regions (it is associated with transcriptional activation) other modifications at H3K4 such as monomethyl is present predominantly at enhancer sequences. Specific marks have been shown to be associated with the activation (H3K9Me1, H3K27Me1, and H4K20Me1) or repression (H3K9Me2 and Me3, H3K27Me2 and Me3, and H4K20Me2 and Me3) of genes. Monomethylation of H4 at K20, catalyzed by SET8, is essential to genome replication and stability. Multiple DNA breaks are associated with demethylation at this site, resulting in activation of p53 to avoid mitosis and aberrant chromosomal activity. In mammalian stem cells, Xist expression blocks the formation of H4K20me1, which is one of the first examples of a direct connection between chromatin and stem cell differentiation.

Histone H3 5CF, 21-44 Control Peptide is suitable for use in ELISA, Western Blot, Dot blot, PCA, and other assays. Control peptide should be used at 1.0 µg per 1.0 µl of antiserum in per assay. Specific conditions for reactivity should be optimized by the end user.