LED Dual Gooseneck Spotlight Illuminator, Aven Tools
Catalog # 76299-818
Supplier: Aven Tools
Specifications
- Description:LED Dual Gooseneck Spotlight
- Cat. No.:76299-818
Specifications
About this item
Ideal external light source, offering versatile and adjustable illumination at any angle.
- Provides bright, even lighting
- 6500K cool light LED illuminator
- Adjustment knobs for each light
Aven’s LED dual gooseneck spotlight illuminator gives users the ability to inspect at any angle and at any working distance. Each flexible gooseneck arm can be swiveled and bent to the desired position and stay in place.