Distilling Adapter, Iowa State, HGF GLASS INC
Catalog # 20203-004
Supplier: HGF Glass
Specifications
- Description:Iowa state distilling adapter with inclined plate, 15.2×15.2 cm
- Cat. No.:20203-004
- Supplier no.:210200-1
Specifications
About this item
Distilling adapter is a component of Iowa state distillation unit which have been used for determinations in the citrus industry but also applicable for other distilling procedures.