About this item
The GloMax® navigator is an easy-to-use microplate luminometer integrated with Promega chemistries for superior luminescence assay performance. It is also possible to interpret results using integrated data analysis software.
- Developed and optimized with Promega cell and gene reporter assays
- Highest instrument sensitivity to measure the lowest assay signals
- Low cross-talk between wells gives you better, more usable results
- Over 30 preloaded assay protocols
- Plate reading luminometer for convenient detection of bioluminescence
- Top-reading, head-on photon counting photomultiplier tube detector
GloMax® navigator provides superior sensitivity and dynamic range for both strong and weak bioluminescence experimental samples as well as seamless integration with promega bioluminescence assays, including gene reporter, cell-based and biochemical assays. The microplate luminometer features optional dual injectors to measure both glow- and flash-based luminescent reactions in 96-well plates.
The luminometer is operated by an integrated tablet PC, which provides quick and easy navigation through the control options. Exporting your results is simple with a variety of options, including export to your local data network, USB flash drive and cloud-based storage location.
*Height of the instrument will be 18" with tablet and PC mounted.
Specifications
- Catalog No:
- 76201-022
- 76201-020
- Detector:
- PMT
- PMT