Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant EIF4E (from E. coli)
Human Recombinant EIF4E (from E. coli)
Certificates
Human Recombinant EIF4E (from E. coli)

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
  • Protein Synonyms:
    Eukaryotic translation initiation factor 4E|eIF-4F 25 kDa subunit|EIF4EL1|EIF4E1|CBP|MGC111573|EIF4F|mRNA cap-binding protein
  • Protein/Peptide Name:
    EIF4E
  • Purity:
    >85%
  • Formulation:
    Liquid (sterile filtered)
  • Shipping Temperature:
    Dry Ice

Specifications