Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant JUN (from E. coli)
Human Recombinant JUN (from E. coli)
Certificates
Human Recombinant JUN (from E. coli)

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
  • Protein Synonyms:
    Transcription factor AP-1|V-jun avian sarcoma virus 17 oncogene homolog|AP1|Proto-oncogene c-Jun|p40|Activator protein 1
  • Protein/Peptide Name:
    JUN
  • Purity:
    >50%
  • Formulation:
    Liquid (sterile filtered)
  • Shipping Temperature:
    Dry Ice

Specifications