Human Recombinant BCKDK (from Baculovirus (Sf9 Insect cells))
BCKDK or branched chain alpha-keto acid dehydrogenase kinase is an important regulator of the valine, leucine, and isoleucine catabolic pathways. BCKDK is found in the mitochondrion where it phosphorylates and inactivates BCKD. The branched-chain alpha-keto dehydrogenase complex (BCKDH) catalyzes the rate-limiting step in the oxidation of branched-chain amino acids and BCKDK inactivates the BCKDH complex by catalyzing serine phosphorylation of the E1-alpha subunit (1). BCKDH kinase catalyzes the serine phosphorylation of the E1- alpha subunit of BCKDH (2). BCKDK Protein is ideal for investigators involved in Singaling Proteins, Cellular Proteins, and Metabolic Disorder research.
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Sf9 Insect cells)
- Species:Human
- Storage Conditions:Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
- Protein Synonyms:[BCKDKD|BCKD-kinase|BCKDHKIN|3-methyl-2-oxobutanoate dehydrogenase [lipoamide]] kinase|mitochondrial|Branched-chain alpha-ketoacid dehydrogenase kinase|BDK
- Protein/Peptide Name:BCKDK
- Purity:>95%
- Formulation:Liquid (sterile filtered)
- Shipping Temperature:Dry Ice
Frequently Bought Together