Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant ANXA5 (from E. coli)
Human Recombinant ANXA5 (from E. coli)
Certificates
Human Recombinant ANXA5 (from E. coli)

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
  • Protein Synonyms:
    Vascular anticoagulant-alpha|Endonexin II|Calphobindin I|ANXA5|Annexin-5|Annexin V|Anchorin CII|PAP-I|Thromboplastin inhibitor|ENX2|ANX5|CBP-I|VAC-alpha|PP4|Lipocortin V|Annexin A5|Placental anticoagulant protein 4
  • Protein/Peptide Name:
    ANXA5
  • Purity:
    >95%
  • Formulation:
    Liquid (sterile filtered)
  • Shipping Temperature:
    Dry Ice

Specifications