Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant EIF4EBP1 (from E. coli)
 
Human Recombinant EIF4EBP1 (from E. coli)

 

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
  • Protein Synonyms:
    Phosphorylated heat- and acid-stable protein regulated by insulin 1|PHAS-I|EIF4EBP1|BP-1|MGC4316|eIF4E-binding protein 1|Eukaryotic translation initiation factor 4E-binding protein 1|4E-BP1
  • Protein/Peptide Name:
    EIF4EBP1
  • Purity:
    >85%
  • Formulation:
    Liquid (sterile filtered)
  • Shipping Temperature:
    Dry Ice