Specifications
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Human
- Size:10 µg
- Storage Conditions:Store vial at -70 °C prior to use. Thaw only prior to immediate use. Maintain sterility. This product DOES NOT contain preservative. DO NOT VORTEX. For long term storage we recommend adding a carrier protein such as HSA or BSA to 0.1% (i.e. 1.0 mg/mL) . For best results aliquot contents and freeze at –20 °C or colder. Avoid cycles of freezing and thawing. Centrifuge vial before each opening to dislodge contents from the cap and to clarify if contents are not clear after standing at room temperature.
- Gene ID:P31749
- Protein Synonyms:RAC|negative control|PKB|AKT|AKT double mutant|PKB alpha|AKT control
- Protein/Peptide Name:AKT1
- Purity:>90%
- Formulation:Liquid
- Shipping Temperature:Dry Ice
- Cat. No.:10801-850
- Supplier no.:009-001-P22
Specifications
About this item
AKT1 is a component of the PI-3 kinase pathway and is activated by phosphorylation at Ser 473 and Thr 308. AKT is a cytoplasmic protein also known as Protein Kinase B (PKB) and RAC (Related to A and C kinases). AKT is a key regulator of many signal transduction pathways, and it exhibits tight control over cell proliferation and cell viability. Overexpression or inappropriate activation of AKT is noted in many types of cancer. AKT mediates many of the downstream events of PI 3-kinase (a lipid kinase activated by growth factors, cytokines and insulin). PI 3-kinase recruits AKT to the membrane, where it is activated by PDK1 phosphorylation. Once phosphorylated, AKT dissociates from the membrane and phosphorylates targets in the cytoplasm and the cell nucleus. AKT has two main roles: (i) inhibition of apoptosis; (ii) promotion of proliferation. AKT1 mutant (T308 / S473A) recombinant protein is ideal for investigators involved in Cell Signaling, Neuroscience and Signal Transduction research.