Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant AKT1 (from Baculovirus (Insect cells))
Human Recombinant AKT1 (from Baculovirus (Insect cells))
Catalog # 10801-846
CAS Number:  
Human Recombinant AKT1 (from Baculovirus (Insect cells))
Catalog # 10801-846
Supplier Number:  009-001-I51
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    Baculovirus (Insect cells)
  • Species:
    Human
  • Size:
    10 µg
  • Storage Conditions:
    Store vial at -70 °C prior to use. Thaw only prior to immediate use. Maintain sterility. This product DOES NOT contain preservative. DO NOT VORTEX. For long term storage we recommend adding a carrier protein such as HSA or BSA to 0.1% (i.e. 1.0 mg/mL) . For best results aliquot contents and freeze at –20 °C or colder. Avoid cycles of freezing and thawing. Centrifuge vial before each opening to dislodge contents from the cap and to clarify if contents are not clear after standing at room temperature.
  • Gene ID:
    P31749
  • Protein Synonyms:
    RAC|PRKBA|PKB|AKT|PKB alpha
  • Protein/Peptide Name:
    AKT1
  • Purity:
    >90%
  • Formulation:
    Liquid
  • Shipping Temperature:
    Dry Ice
  • Cat. No.:
    10801-846
  • Supplier no.:
    009-001-I51

Specifications

About this item

AKT1 is a component of the PI-3 kinase pathway and is activated by phosphorylation at Ser 473 and Thr 308. AKT is a cytoplasmic protein also known as Protein Kinase B (PKB) and RAC (Related to A and C kinases). AKT is a key regulator of many signal transduction pathways, and it exhibits tight control over cell proliferation and cell viability. Overexpression or inappropriate activation of AKT is noted in many types of cancer. AKT mediates many of the downstream events of PI 3-kinase (a lipid kinase activated by growth factors, cytokines and insulin). PI 3-kinase recruits AKT to the membrane, where it is activated by PDK1 phosphorylation. Once phosphorylated, AKT dissociates from the membrane and phosphorylates targets in the cytoplasm and the cell nucleus. AKT has two main roles: (i) inhibition of apoptosis; (ii) promotion of proliferation. AKT1 recombinant protein is ideal for investigators involved in Cell Signaling, Neuroscience and Signal Transduction research.