Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant TNFSF11 (from E. coli)
Human Recombinant TNFSF11 (from E. coli)
  RL009001C22
 :  
Human Recombinant TNFSF11 (from E. coli)
  RL009001C22
 :  009-001-C22
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    10 µg
  • Storage Conditions:
    Store vial at 4 °C prior to restoration. Dilute only prior to immediate use. Maintain sterility. This product DOES NOT contain preservative. DO NOT VORTEX. We recommend adding a carrier protein such as HSA or BSA to 0.1% (i.e. 1.0 mg/mL) . For best results aliquot contents and freeze at –20 °C or colder. Avoid cycles of freezing and thawing. Centrifuge vial before each opening to dislodge contents from the cap and to clarify if contents are not clear after standing at room temperature.
  • Endotoxin Content:
    Measured by kinetic LAL analysis and is typically ≤ 1 EU/μg protein.
  • Biological Activity:
    RANK ligand is fully biologically active when compared to standard. The activity is determined by activity of RAW-Blue™ cells using a concentration of 50ng/ml and by dose dependent stimulation of IL-8 production in human PBMC.
  • Gene ID:
    O14788
  • Reconstitution Instructions:
    10µl (10-100µl)
  • Protein Synonyms:
    Osteoprotegerin ligand|OPGL|ODF|TRANCE|Rank Ligand|Receptor activator of nuclear factor kappa-B ligand|CD254|TNF-related activation-induced cytokine|Osteoclast differentiation factor|Tumor necrosis factor ligand superfamily member 11
  • Protein/Peptide Name:
    TNFSF11
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    RL009001C22
  • Supplier no.:
    009-001-C22

 

 

The secreted cytokine RANKL (Receptor Activator of Nuclear factor kappa-B Ligand) is critically involved in osteoclastic differentiation and activation and in the regulation of specific immunity. RANKL exists as a homotrimer, is glycosylated, and occurs in 3 forms: cell-bound RANKL, which is expressed by osteoblast lineage cells, soluble RANKL (sRANKL), which is expressed by activated T lymphocytes, and a truncated ectodomain form derived from the cell-bound RANK Ligand, which is enzymatically processed by TACE (TNF-alpha converting enzyme (TACE; ADAM-17)). All three forms stimulate their specific receptor, RANK, which is located on osteoclastic and dendritic cells. RANKL binds to TNFRSF11B/OPG and to TNFRSF11A/RANK. RANKL augments the ability of dendritic cells to stimulate naive T-cell proliferation. It may be an important regulator of interactions between T-cells and dendritic cells and may play a role in the regulation of the T-cell-dependent immune response. It may also play an important role in enhanced bone-resorption in humoral hypercalcemia of malignancy. Expression of RANKL is highest in the peripheral lymph nodes, weak in spleen, peripheral blood leukocytes, bone marrow, heart, placenta, skeletal muscle, stomach and thyroid and is up-regulated by T-cell receptor stimulation. RANKL is secreted in the soluble form. Defects in TNFSF11 cause osteopetrosis. Recombinant human soluble RANKL produced in E.coli is a single, non-glycosylated polypeptide chain containing 176 amino acids and having a molecular mass of 20,006 Daltons.