Order Entry
Puerto Rico
ContactUsLinkComponent
Polycarbonate Bottles, BECKMAN COULTER
Polycarbonate Bottles, BECKMAN COULTER
Catalog # BK366751
Supplier:  Beckman Coulter
Product available on GSA Advantage®
undefined
Polycarbonate Bottles, BECKMAN COULTER
Catalog # BK366751
Supplier:  Beckman Coulter
Supplier Number:  366751
Product available on GSA Advantage®

Specifications

  • Capacity:
    1000 ml
  • Description:
    Polycarbonate bottle
  • Size:
    95×191 mm
  • Cat. No.:
    BK366751
  • Max. rcf:
    16800×g
  • Supplier no.:
    366751

Specifications

About this item

Polycarbonate bottle for use in high performance and high capacity rotors.