9-Piece Organic Chemistry Glassware Kit, United Scientific Supplies
Catalog # 76293-308
Supplier: United Scientific Supplies
Specifications
- Description:9-Piece Organic Chemistry Glassware Kit
- Environmentally Preferable:
- Cat. No.:76293-308
Specifications
About this item
This 9-piece standard set is designed to cover essential requirements for organic chemistry and is suitable for preparations up to 30 g.
- All glassware is jointed
- Supplied in a sturdy, form fitting case
- All Joints are 14/23
This starter set is designed for organic chemistry labs and is suitable for small-scale procedures up to 30g. All of the ground glass components in this set have a joint size of 14/23. Includes a sturdy storage box with foam insert.
Delivery: Kit includes: Pear shaped flask (50 ml), still head, liebig condenser, screw cap adapter, receiver adapter, air leak/steam inlet tube, dropping funnel (50 ml with rotoflo tap), stopper and thermometer.