Cast Aluminum Clamp Holder, United Scientific Supplies
76293-416
- Description:Cast Aluminum Clamp Holder
- Cat. No.:76293-416
Easily connect items to a support rod.
- Easy to adjust thumbscrews
- Fits rods up to 19 mm
Cast iron body with brass thumb screws.