Order Entry
Puerto Rico
ContactUsLinkComponent
ChIP DNA Clean and Concentrator™ Kits, Zymo Research
ChIP DNA Clean and Concentrator™ Kits, Zymo Research
Supplier:  Zymo Research
Certificates
undefined
ChIP DNA Clean and Concentrator™ Kits, Zymo Research
Supplier:  Zymo Research

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Immunoprecipitates
  • Target molecule:
    DNA
  • Description:
    ChIP DNA Clean Concentrator Kit

Specifications

Related Information

Frequently Bought Together