Order Entry
United States
ContactUsLinkComponent
DNA/RNA Shield™ Fecal Collection Tubes
DNA/RNA Shield™ Fecal Collection Tubes
Catalog # 76020-422
Supplier:  Zymo Research
undefined
DNA/RNA Shield™ Fecal Collection Tubes
Catalog # 76020-422
Supplier:  Zymo Research
Supplier Number:  R1101

Specifications

  • Description:
    DNA/RNA Shield™ fecal collection tubes
  • Dimensions:
    20×76 mm
  • Size:
    10 pack
  • Environmentally Preferable:
  • Cat. No.:
    76020-422
  • Additive:
    with DNA/RNA shield (9 ml)

Specifications

About this item

The DNA/RNA Shield™ fecal collection tube is powered by the DNA/RNA shield sample stabilization reagent, which takes a microbial snapshot of a sample, inactivates viruses, and makes samples safe and transport ready. Samples are stable at ambient temperature and can be frozen for longer-term storage.

  • Spoon attached to screwcap
  • RNA: Ambient temperature (4 to 25 °C) >1 month
  • DNA: Ambient temperature (4 to 25 °C) >2 years
  • DNA and RNA: Frozen (<–20 °C) indefinitely

Samples collected are ready for all sensitive downstream analyses (microbiomic, next-generation sequencing, qPCR, microarray and more.

DNA/RNA transport medium for stool samples. 1 g or 1 ml stool, soil, environmental samples can be collected.