10X PBS with 0.5% Tween 20, Teknova
Catalog # 101320-452
Supplier: Teknova
CAS Number:
Specifications
- Description:10X PBS with 0.5% Tween 20
- Size:1 l
- Cat. No.:101320-452
- pH:7.4 (1X)
- Supplier no.:P0197
Specifications
About this item
Tween 20 is a frequently used member of the polysorbate family of detergents.
- Sterile
- Shipping at ambient conditions
- Store at room temperature
Contains 1.37 M sodium chloride (NaCl); 27 mM potassium chloride (KCl); 79.4 mM disodium phosphate (Na2PO4); 20.6 mM monopotassium phosphate (KH2PO4) and 0.5% (v/v) Tween-20 (polysorbate 20).