Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant BMP-4 (from CHO cells)
Human Recombinant BMP-4 (from CHO cells)
Certificates
undefined
Human Recombinant BMP-4 (from CHO cells)

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: Alkaline phosphatase activity induced in ATDC-5 cells, ED50 ≤15 ng/ml, bioactivity ≥6.7×10⁴ units/mg
  • Gene ID:
    P12644
  • Reconstitution Instructions:
    Sterile 10 mM HCl at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein Synonyms:
    BMP-2B
  • Protein/Peptide Name:
    BMP-4
  • Purity:
    ≥95%
  • Molecular Weight:
    13.1/26.2 kDa
  • Sequence:
    SPKHHSQRAR KKNKNCRRHS LYVDFSDVGW NDWIVAPPGY QAFYCHGDCP FPLADHLNST NHAI VQT LVN SVNSSIPKAC CVPTELSAIS MLYLDEYDKV VLKNYQEMVV EGCGCR
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay

Specifications