Anti-CD3D and CD3E Rabbit Monoclonal Antibody [clone: 301] PerCP (Peridinin-Chlorophyll Protein Complex)
Supplier: Sino Biological
Certificates
About this item
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD3D and CD3E (rh CD3D and CD3E; Catalog#CT026-H0323H; P04234-1 and P07766-1; Met1-Ala105 and Met1-Asp126) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Type: Primary
Antigen: CD3D/CD3E
Clonality: Monoclonal
Clone: Clone: 301
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Specifications
- Catalog No:
- 103632-482
- 103630-286
- Antigen Symbol:
- CD3D & CD3E
- CD3D & CD3E
- Antigen Name:
- CD3 epsilon & CD3 delta Heterodimer Protein
- CD3 epsilon & CD3 delta Heterodimer Protein
- Antigen Synonyms:
- T3d
- T3d
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PerCP (Peridinin-Chlorophyll Protein Complex)
- PerCP (Peridinin-Chlorophyll Protein Complex)
- Clone:
- 301
- 301
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Flow Cytometry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- Shipping Temperature:
- Ambient
- Ambient