Anti-CD3D and CD3E Rabbit Monoclonal Antibody [clone: 301]
Supplier: Sino Biological
Certificates
About this item
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD3D and CD3E Heterodimer (Catalog#CT026-H0323H; P04234-1 and P07766-1; Met1-Ala105 and Met1-Asp126).
Type: Primary
Antigen: CD3D/CD3E
Clonality: Monoclonal
Clone: 301
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Specifications
- Catalog No:
- 103662-198
- Antigen Symbol:
- CD3D & CD3E
- Antigen Name:
- CD3 epsilon & CD3 delta Heterodimer Protein
- Antigen Synonyms:
- T3d
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 301
- Reactivity:
- Cynomolgus monkey, Human
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Flow Cytometry:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm Filtered solution in PBS
- Shipping Temperature:
- Ambient