Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD4 Mouse Monoclonal Antibody [clone: 02] PE (Phycoerythrin)
Anti-CD4 Mouse Monoclonal Antibody [clone: 02] PE (Phycoerythrin)
Supplier:  Sino Biological
Certificates
undefined
Anti-CD4 Mouse Monoclonal Antibody [clone: 02] PE (Phycoerythrin)
Supplier:  Sino Biological

Specifications

  • Catalog No:
  • 103632-542
  • 103630-210
  • Antigen Symbol:
  • CD4
  • CD4
  • Antigen Name:
  • CD4 molecule
  • CD4 molecule
  • Antigen Synonyms:
  • CD4mut
  • CD4mut
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • 02
  • 02
  • Reactivity:
  • Ferret
  • Ferret
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • Flow Cytometry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Storage Buffer:
  • PBS solution containing 0.5% BSA and 0.03% ProClin300
  • PBS solution containing 0.5% BSA and 0.03% ProClin300
  • Shipping Temperature:
  • Ambient
  • Ambient

Specifications

Related Information

Frequently Bought Together