Anti-CD4 Mouse Monoclonal Antibody [clone: 02] PerCP (Peridinin-Chlorophyll Protein Complex)
Supplier: Sino Biological
Certificates
About this item
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Ferret CD4 (rFerret CD4; Catalog#60003-F08H; ABS50090.1; Met1-Leu401) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Type: Primary
Antigen: Anti-CD4 Antibody (PerCP)
Clonality: Monoclonal
Clone: Clone: 02
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Ferret
Specifications
- Catalog No:
- 103630-206
- 103632-386
- Antigen Symbol:
- CD4
- CD4
- Antigen Name:
- CD4 molecule
- CD4 molecule
- Antigen Synonyms:
- CD4mut
- CD4mut
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PerCP (Peridinin-Chlorophyll Protein Complex)
- PerCP (Peridinin-Chlorophyll Protein Complex)
- Clone:
- 02
- 02
- Reactivity:
- Ferret
- Ferret
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Flow Cytometry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- Shipping Temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together