Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Anti-CDCP1 Rabbit Polyclonal Antibody

Anti-CDCP1 Rabbit Polyclonal Antibody

Anti-CDCP1 Rabbit Polyclonal Antibody
 
Anti-CDCP1 Rabbit Polyclonal Antibody

 

 

Produced in rabbits immunized with purified, recombinant Mouse CDCP1 (rM CDCP1; Catalog#51110-M08H; Q5U462; Met1-Leu666). CDCP1 specific IgG was purified by Mouse CDCP1 affinity chromatography.

Type: Primary
Antigen: CDCP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

 

 

  • Catalog No:
  • 103596-148
  • 103659-958
  • 103659-960
  • Antigen Symbol:
  • CDCP1
  • CDCP1
  • CDCP1
  • Antigen Name:
  • CUB domain containing protein 1
  • CUB domain containing protein 1
  • CUB domain containing protein 1
  • Antigen Synonyms:
  • TRASK|SIMA135|CD318
  • TRASK|SIMA135|CD318
  • TRASK|SIMA135|CD318
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Mouse
  • Mouse
  • Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Liquid
  • Storage Buffer:
  • 0.2 μm Filtered solution in PBS
  • 0.2 μm Filtered solution in PBS
  • 0.2 μm Filtered solution in PBS
  • Shipping Temperature:
  • Ambient
  • Ambient
  • Ambient

 

Product Family Options

 

  • Size
     
     
  • 50 µl
     
     : -
     
     
     
    Specifications
    Cat. No.
    103596-148
    Antigen Symbol
    CDCP1
    Antigen Name
    CUB domain containing protein 1
    Antigen Synonyms
    TRASK|SIMA135|CD318
    Antibody Type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Mouse
    Host
    Rabbit
    Isotype
    IgG
    ELISA
    Yes
    ImmunoChemistry
    Yes
    Form
    Liquid
    Storage Buffer
    0.2 μm Filtered solution in PBS
    Shipping Temperature
    Ambient
  • 100 µl
       
       
    Each (100µG)
    Each (100µG) - $274.50
    $274.50
    Each (100µG)
       $274.50
    Minimum order quantity is 1

     : 51110-RP02-100
     
     
     
    Specifications
    Cat. No.
    103659-958
    Antigen Symbol
    CDCP1
    Antigen Name
    CUB domain containing protein 1
    Antigen Synonyms
    TRASK|SIMA135|CD318
    Antibody Type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Mouse
    Host
    Rabbit
    Isotype
    IgG
    ELISA
    Yes
    ImmunoChemistry
    Yes
    Form
    Liquid
    Storage Buffer
    0.2 μm Filtered solution in PBS
    Shipping Temperature
    Ambient
  • 200 µl
     
     : -
     
     
     
    Specifications
    Cat. No.
    103659-960
    Antigen Symbol
    CDCP1
    Antigen Name
    CUB domain containing protein 1
    Antigen Synonyms
    TRASK|SIMA135|CD318
    Antibody Type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Mouse
    Host
    Rabbit
    Isotype
    IgG
    ELISA
    Yes
    ImmunoChemistry
    Yes
    Form
    Liquid
    Storage Buffer
    0.2 μm Filtered solution in PBS
    Shipping Temperature
    Ambient
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.