Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-PROCR Rabbit Monoclonal Antibody [clone:101]
Anti-PROCR Rabbit Monoclonal Antibody [clone:101]
Supplier:  Sino Biological
Certificates
Anti-PROCR Rabbit Monoclonal Antibody [clone:101]
Supplier:  Sino Biological

Specifications

  • Catalog No:
  • 103659-622
  • Antigen Symbol:
  • PROCR
  • Antigen Name:
  • Protein C receptor
  • Antigen Synonyms:
  • EPCR|CCCA|CCD41
  • Antibody Type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • 101
  • Reactivity:
  • Mouse
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • ELISA:
  • Yes
  • Form:
  • Liquid
  • Storage Buffer:
  • 0.2 μm filtered solution in PBS
  • Shipping Temperature:
  • Ambient

Specifications