Anti-EphA2 Rabbit Monoclonal Antibody [clone:301] FITC (Fluorescein Isothiocyanate)
Supplier: Sino Biological
Certificates
About this item
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EphA2 / Eph Receptor A2 (rM EphA2 / Eph Receptor A2; Catalog#50586-M08H; NP_034269.2; Met1-Asn535) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Type: Primary
Antigen: EphA2
Clonality: Monoclonal
Clone: Clone: 301
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 103630-112
- 103632-276
- Antigen Symbol:
- EphA2
- EphA2
- Antigen Name:
- EPH receptor A2
- EPH receptor A2
- Antigen Synonyms:
- CTPA|ARCC2|ECK|CTRCT6|CTPP1
- CTPA|ARCC2|ECK|CTRCT6|CTPP1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- 301
- 301
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Flow Cytometry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- Shipping Temperature:
- Ambient
- Ambient