Anti-METAP2 Rabbit Polyclonal Antibody
Supplier: Sino Biological
Certificates
About this item
Produced in rabbits immunized with purified, recombinant Mouse METAP2 / MAP2 (rM METAP2 / MAP2; Catalog#50528-M08B; O08663; Ala2-Tyr478). METAP2 / MAP2 specific IgG was purified by Mouse METAP2 / MAP2 affinity chromatography.
Type: Primary
Antigen: MAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 103658-274
- Antigen Symbol:
- METAP2
- Antigen Name:
- Methionyl aminopeptidase 2
- Antigen Synonyms:
- MNPEP|MAP2|p67eIF2|p67
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Mouse
- Host:
- Rabbit
- Isotype:
- IgG
- Western Blot:
- Yes
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- ImmunoPrecipitation:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient