Anti-SPARC Rabbit Polyclonal Antibody
Supplier: Sino Biological
Certificates
About this item
Produced in rabbits immunized with purified, recombinant Mouse Osteonectin / SPARC (rM Osteonectin / SPARC; Catalog#50494-M08H; NP_033268.1; Met 1-Ile 302). Osteonectin / SPARC specific IgG was purified by Mouse Osteonectin / SPARC affinity chromatography.
Type: Primary
Antigen: SPARC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 103659-350
- Antigen Symbol:
- SPARC
- Antigen Name:
- Secreted protein acidic and cysteine rich
- Antigen Synonyms:
- ON|SPARC|BM-40
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Mouse
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Specifications
Frequently Bought Together