Anti-ECM1 Rabbit Monoclonal Antibody [clone:038]
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ECM1 (rM ECM1; Catalog#50331-M08H; Q61508-1; Met 1-Glu 559).
Type: Primary
Antigen: ECM1
Clonality: Monoclonal
Clone: 038
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
- Catalog No:
- 103657-794
- Antigen Symbol:
- ECM1
- Antigen Name:
- Extracellular matrix protein 1
- Antigen Synonyms:
- URBWD
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 038
- Reactivity:
- Mouse
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient